Gene Transcripts

Gene Symbol
Arhgef26
Entrez Gene ID
310460
Gene Name
Rho guanine nucleotide exchange factor (GEF) 26
Gene Aliases
RGD1562559, Sgef
Chromosome Location
Chr. 2: 27107276 - 27236336 on Build Rnor_6.0
UniGene ID
Rn.7792
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_227201.8 XP_227201.4 14 3088
GenBank mRNA FQ218213.1 - 13 1733

View Full Product Details