Gene Transcripts

Gene Symbol
Gtpbp10
Entrez Gene ID
312054
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
Cldn12
Chromosome Location
Chr. 4: 25635765 - 25653283 on Build Rnor_6.0
UniGene ID
Rn.139851
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100815.1 NP_001094285.1 10 1023
GenBank mRNA CA508019.1 - 1 645

View Full Product Details