Gene Transcripts

Gene Symbol
Vit
Entrez Gene ID
313831
Gene Name
vitrin
Gene Aliases
RGD1564128
Chromosome Location
Chr. 6: 1115645 - 1208888 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764427.1 XP_008762649.1 5 453

View Full Product Details