Gene Transcripts

Gene Symbol
Unc79
Entrez Gene ID
314401
Gene Name
unc-79 homolog (C. elegans)
Gene Aliases
RGD1311117
Chromosome Location
Chr. 6: 126821503 - 127069026 on Build Rnor_6.0
UniGene ID
Rn.43444
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271253.1 NP_001258182.1 31 5581
XM_006240501.2 XP_006240563.1 32 5870

View Full Product Details