Gene Transcripts

Gene Symbol
Tbc1d10c
Entrez Gene ID
361697
Gene Name
TBC1 domain family, member 10C
Gene Aliases
RGD1311490
Chromosome Location
Chr. 1: 219431094 - 219438839 on Build Rnor_6.0
UniGene ID
Rn.61004
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081980.1 NP_001075449.1 2 75
XM_006230840.2 XP_006230902.1 2 128
GenBank mRNA BC160813.1 - 2 86
FQ226277.1 - 2 86

View Full Product Details