Gene Transcripts

Gene Symbol
Nrxn1
Entrez Gene ID
60391
Gene Name
neurexin 1
Gene Aliases
Nrxn1b
Chromosome Location
Chr. 6: 13886757 - 13908139 on Build Rnor_6.0
UniGene ID
Rn.54924
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021767.2 NP_068535.2 2787
GenBank mRNA M96374.1 - 2787

View Full Product Details