Gene Transcripts

Gene Symbol
Tas2r13
Entrez Gene ID
78984
Gene Name
taste receptor, type 2, member 13
Gene Aliases
T2R8
Chromosome Location
Chr. 4: 166958466 - 166959410 on Build Rnor_6.0
UniGene ID
Rn.48787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023998.1 NP_076488.1 1 518
GenBank mRNA AF227145.1 - 1 518

View Full Product Details