Gene Transcripts

Gene Symbol
Tek
Entrez Gene ID
89804
Gene Name
TEK tyrosine kinase, endothelial
Gene Aliases
Tie-2, Tie2
Chromosome Location
Chr. 5: 113725717 - 113852799 on Build Rnor_6.0
UniGene ID
Rn.9159
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105737.1 NP_001099207.1 21 3445
XM_008763910.1 XP_008762132.1 21 3265

View Full Product Details