Gene Transcripts

Gene Symbol
Kirrel2
Entrez Gene ID
100359836
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 1: 88909838 - 88920834 on Build Rnor_6.0
UniGene ID
Rn.234307
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271398.1 NP_001258327.1 11 2097
XM_008759099.1 XP_008757321.1 12 2053

View Full Product Details