Gene Transcripts

Gene Symbol
Mmp1
Entrez Gene ID
300339
Gene Name
matrix metallopeptidase 1 (interstitial collagenase)
Gene Aliases
Mmp1a
Chromosome Location
Chr. 8: 5703206 - 5723593 on Build Rnor_6.0
UniGene ID
Rn.79007
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134530.1 NP_001128002.1 10 1389
GenBank mRNA BC169019.1 - 10 1389

View Full Product Details