Gene Transcripts

Gene Symbol
Ccdc12
Entrez Gene ID
363151
Gene Name
coiled-coil domain containing 12
Gene Aliases
RGD1306081
Chromosome Location
Chr. 8: 118925681 - 118977038 on Build Rnor_6.0
UniGene ID
Rn.11535
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108783.1 NP_001102253.1 1 112
XM_006243970.2 XP_006244032.1 2 736

View Full Product Details