Gene Transcripts

Gene Symbol
Prl7b1
Entrez Gene ID
266804
Gene Name
prolactin family 7, subfamily b, member 1
Gene Aliases
Prlpn
Chromosome Location
Chr. 17: 39153431 - 39161644 on Build Rnor_6.0
UniGene ID
Rn.79989
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153738.1 NP_714960.1 5 445
GenBank mRNA AF525159.1 - 5 445

View Full Product Details