Gene Transcripts

Gene Symbol
Csn1s2b
Entrez Gene ID
286759
Gene Name
casein alpha s2-like B
Gene Aliases
Csnd
Chromosome Location
Chr. 14: 21899954 - 21916068 on Build Rnor_6.0
UniGene ID
Rn.91144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173106.1 NP_775129.1 232
GenBank mRNA AY154894.1 - 232

View Full Product Details