Gene Transcripts

Gene Symbol
Zdhhc25
Entrez Gene ID
300323
Gene Name
zinc finger, DHHC-type containing 25
Gene Aliases
DHHC26
Chromosome Location
Chr. 7: 129595192 - 129596424 on Build Rnor_6.0
UniGene ID
Rn.124773
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039333.1 NP_001034422.1 1 331
GenBank mRNA AY886541.1 - 1 331

View Full Product Details