Gene Transcripts

Gene Symbol
Nfu1
Entrez Gene ID
297416
Gene Name
NFU1 iron-sulfur cluster scaffold
Gene Aliases
Hirip5
Chromosome Location
Chr. 4: 118814284 - 118834955 on Build Rnor_6.0
UniGene ID
Rn.161843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106606.2 NP_001100076.2 7 576
XM_008763063.1 XP_008761285.1 7 809
GenBank mRNA FM064033.1 - 6 402

View Full Product Details