Gene Transcripts

Gene Symbol
Srbd1
Entrez Gene ID
301665
Gene Name
S1 RNA binding domain 1
Gene Aliases
RGD1562788
Chromosome Location
Chr. 6: 9266559 - 9459219 on Build Rnor_6.0
UniGene ID
Rn.105705
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239708.2 XP_006239770.1 14 2006
XM_237474.7 XP_237474.5 14 1875
GenBank mRNA FQ212138.1 - 11 1476

View Full Product Details