Gene Transcripts

Gene Symbol
Slc5a10
Entrez Gene ID
303205
Gene Name
solute carrier family 5 (sodium/sugar cotransporter), member 10
Gene Aliases
Chromosome Location
Chr. 10: 47949777 - 47997803 on Build Rnor_6.0
UniGene ID
Rn.218380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107007.1 NP_001100477.1 5 409
XM_008767820.1 XP_008766042.1 5 609
GenBank mRNA BC161867.1 - 5 602

View Full Product Details