Gene Transcripts

Gene Symbol
Pnmal1
Entrez Gene ID
361515
Gene Name
paraneoplastic Ma antigen family-like 1
Gene Aliases
RGD1310803
Chromosome Location
Chr. 1: 78883255 - 78896551 on Build Rnor_6.0
UniGene ID
Rn.66585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108473.1 NP_001101943.1 2 192
XM_006228440.2 XP_006228502.1 3 847
XM_008758951.1 XP_008757173.1 4 563

View Full Product Details