Gene Transcripts

Gene Symbol
Pram1
Entrez Gene ID
362848
Gene Name
PML-RARA regulated adaptor molecule 1
Gene Aliases
RGD1564330
Chromosome Location
Chr. 7: 18507896 - 18516299 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241132.2 XP_006241194.1 3, 4 1850
XM_008765204.1 XP_008763426.1 3, 4 1850

View Full Product Details