Gene Transcripts

Gene Symbol
Ffar3
Entrez Gene ID
365228
Gene Name
free fatty acid receptor 3
Gene Aliases
Gpr41
Chromosome Location
Chr. 1: 89302507 - 89303847 on Build Rnor_6.0
UniGene ID
Rn.122324
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108912.1 NP_001102382.1 2 734
GenBank mRNA BC166522.1 - 2 855

View Full Product Details