Gene Transcripts

Gene Symbol
Clcf1
Entrez Gene ID
365395
Gene Name
cardiotrophin-like cytokine factor 1
Gene Aliases
Bsf3, Clc, NNT-1
Chromosome Location
Chr. 1: 219468866 - 219472445 on Build Rnor_6.0
UniGene ID
Rn.130193
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207615.2 NP_997498.2 2 458
GenBank mRNA AY365006.1 - 2 205
BC098643.1 - 2 458 7123848

View Full Product Details