Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006231418.2 | XP_006231480.1 | 2 | 4095 | |
XM_008760470.1 | XP_008758692.1 | 1 | 4072 | ||
XM_008760471.1 | XP_008758693.1 | 1 | 4075 | ||
XM_008760472.1 | XP_008758694.1 | 1 | 4071 |