Gene Transcripts

Gene Symbol
Krt26
Entrez Gene ID
407758
Gene Name
keratin 26, type I
Gene Aliases
Ka39
Chromosome Location
Chr. 10: 87239722 - 87249121 on Build Rnor_6.0
UniGene ID
Rn.231027
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008823.1 NP_001008823.1 1 197
XM_006247390.2 XP_006247452.1 1 746

View Full Product Details