Gene Transcripts

Gene Symbol
Vom1r68
Entrez Gene ID
494255
Gene Name
vomeronasal 1 receptor 68
Gene Aliases
V1rc1
Chromosome Location
Chr. 4: 87440996 - 87441907 on Build Rnor_6.0
UniGene ID
Rn.129672
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008906.1 NP_001008906.1 1 154
GenBank mRNA AY510275.1 - 1 154

View Full Product Details