Gene Transcripts

Gene Symbol
Slmo2
Entrez Gene ID
494346
Gene Name
slowmo homolog 2 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 3: 172567092 - 172574382 on Build Rnor_6.0
UniGene ID
Rn.185814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009543.1 NP_001009543.1 2 246
GenBank mRNA BC060590.1 - 2 246

View Full Product Details