Gene Transcripts

Gene Symbol
Flrt1
Entrez Gene ID
499308
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
RGD1565152
Chromosome Location
Chr. 1: 222340481 - 222360684 on Build Rnor_6.0
UniGene ID
Rn.169901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109160.1 NP_001102630.1 1 1198
XM_008760167.1 XP_008758389.1 2 4531

View Full Product Details