Gene Transcripts

Gene Symbol
Stra8
Entrez Gene ID
500079
Gene Name
stimulated by retinoic acid 8
Gene Aliases
RGD1562852
Chromosome Location
Chr. 4: 62447724 - 62474780 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236282.2 XP_006236344.1 2 2595
XM_575429.4 XP_575429.2 1 338

View Full Product Details