Gene Transcripts

Gene Symbol
Ly49s5
Entrez Gene ID
503662
Gene Name
Ly49 stimulatory receptor 5
Gene Aliases
Chromosome Location
Chr. 4: 164284910 - 164306500 on Build Rnor_6.0
UniGene ID
Rn.154805
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012749.1 NP_001012767.1 2 226
GenBank mRNA AY649832.1 - 2 226
AY846265.1 - 1 118

View Full Product Details