Gene Transcripts

Gene Symbol
U2af1
Entrez Gene ID
687575
Gene Name
U2 small nuclear RNA auxiliary factor 1
Gene Aliases
Chromosome Location
Chr. 20: 10396652 - 10407557 on Build Rnor_6.0
UniGene ID
Rn.234397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044291.1 NP_001037756.1 6 428
XM_006256228.2 XP_006256290.1 6 445
GenBank mRNA BC105905.1 - 6 428
FQ229339.1 - 6 422

View Full Product Details