Gene Transcripts

Gene Symbol
MAP4K3
Entrez Gene ID
8491
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases
GLK, MAPKKKK3, MEKKK 3, MEKKK3, RAB8IPL1
Chromosome Location
Chr. 2: 39249266 - 39437312 on Build GRCh38
UniGene ID
Hs.655750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270425.1 NP_001257354.1 29 2586
NM_003618.3 NP_003609.2 30 2649
GenBank mRNA AK021553.1 - 2 153
AK291467.1 - 30 2615
AK299889.1 - 22 1678
AK314487.1 - 29 2423
AK316144.1 - 3 229
BC071579.1 - 29 2578 5296205

View Full Product Details