Gene Transcripts

Gene Symbol
Mir17hg
Entrez Gene ID
75957
Gene Name
Mir17 host gene (non-protein coding)
Gene Aliases
5033413D16Rik, Mirhg1, OTTMUSG00000033261
Chromosome Location
Chr. 14: 115044390 - 115046728 on Build GRCm38
UniGene ID
Mm.34859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029382.1 1 439
GenBank mRNA AK053349.1 - 1 332
BQ557174.1 - 1 439

View Full Product Details