Gene Transcripts

Gene Symbol
Ythdf3
Entrez Gene ID
229096
Gene Name
YTH domain family 3
Gene Aliases
9130022A11Rik
Chromosome Location
Chr. 3: 16183005 - 16217037 on Build GRCm38
UniGene ID
Mm.23834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027375.1 3 414
GenBank mRNA AK039176.1 - 3 361
AK053736.1 - 3 333
BC057158.1 - 3 325 5348717


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ121779.1 - 3 316 Chr. 3 - 16183282 - 16189563 [+]
GenBankmRNA BB852061.1 - 3 338 Chr. 3 - 16183268 - 16189513 [+]
GenBankmRNA BB665437.1 - 3 333 Chr. 3 - 16183264 - 16203951 [+]
GenBankmRNA BI659742.1 - 3 323 Chr. 3 - 16183279 - 16203952 [+]
GenBankmRNA BB633007.1 - 3 361 Chr. 3 - 16183236 - 16203921 [+]
GenBankmRNA CJ141260.1 - 3 362 Chr. 3 - 16183236 - 16203924 [+]

View Full Product Details