Gene Transcripts
- Gene Symbol
- Thap6
- Entrez Gene ID
- 381650
- Gene Name
- THAP domain containing 6
- Gene Aliases
- 4930534K21Rik, AI448650
- Chromosome Location
- Chr. 5: 91962882 - 91972137 on Build GRCm38
- UniGene ID
- Mm.159453
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028429.1 |
|
2, 3 |
736 |
|
GenBank mRNA |
AK015967.1 |
- |
3, 4 |
973 |
|
BC089377.1 |
- |
2, 3 |
285 |
|
View Full Product Details