Gene Transcripts

Gene Symbol
Phf21b
Entrez Gene ID
271305
Gene Name
PHD finger protein 21B
Gene Aliases
A730032D07Rik
Chromosome Location
Chr. 15: 84785376 - 84856132 on Build GRCm38
UniGene ID
Mm.101022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_030731.1 8, 9 1291
GenBank mRNA AK046530.1 - 8, 9 1216
BC172145.1 - 8, 9 913 9054815

View Full Product Details