Gene Transcripts
- Gene Symbol
- Scarna13
- Entrez Gene ID
- 100306943
- Gene Name
- small Cajal body-specific RNA 1
- Gene Aliases
- U93
- Chromosome Location
- Chr. 12: 105031075 - 105031349 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028576.1 |
|
1 |
250 |
|
- Gene Symbol
- Mir3069
- Entrez Gene ID
- 100526471
- Gene Name
- microRNA 3069
- Gene Aliases
- mir-3069, mmu-mir-3069
- Chromosome Location
- Chr. 12: 105031077 - 105031141 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037229.1 |
|
1 |
42 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE633860.1 |
- |
1 |
385 |
|
Chr. 12 - 105031013 - 105031485 [-] |
View Full Product Details