Gene Transcripts
- Gene Symbol
- Snora21
- Entrez Gene ID
- 100302498
- Gene Name
- small nucleolar RNA, H/ACA box 21
- Gene Aliases
- MBI-3
- Chromosome Location
- Chr. 11: 97781639 - 97781766 on Build GRCm38
- UniGene ID
- Mm.453536
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028078.1 |
|
1 |
23 |
|
GenBank mRNA |
AF357384.1 |
- |
1 |
23 |
|
View Full Product Details