Gene Transcripts
- Gene Symbol
- Vmn2r-ps11
- Entrez Gene ID
- 319210
- Gene Name
- vomeronasal 2, receptor, pseudogene 11
- Gene Aliases
- 4930518C23Rik
- Chromosome Location
- Chr. 3: 64632863 - 64844743 on Build GRCm38
- UniGene ID
- Mm.246646
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003962.1 |
|
1 |
179 |
|
GenBank mRNA |
AK029734.1 |
- |
1 |
180 |
|
BB615497.1 |
- |
1 |
180 |
|
- Gene Symbol
- Vmn2r7
- Entrez Gene ID
- 319217
- Gene Name
- vomeronasal 2, receptor 7
- Gene Aliases
- 4933425M15Rik
- Chromosome Location
- Chr. 3: 64690660 - 64719602 on Build GRCm38
- UniGene ID
- Mm.246646
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030224.1 |
- |
|
201 |
|
View Full Product Details