Gene Transcripts

Gene Symbol
ZSCAN12P1
Entrez Gene ID
221584
Gene Name
zinc finger and SCAN domain containing 12 pseudogene 1
Gene Aliases
ZNF187p1, ZNF305P1, ZNF96L1, ZNF96P1, ZSCAN12L1, dJ313I6.7
Chromosome Location
Chr. 6: 28090807 - 28095715 on Build GRCh38
UniGene ID
Hs.634723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024063.2 2 3676
GenBank mRNA AK127188.1 - 1 1198
BC040524.1 - 2 3332

View Full Product Details