Gene Transcripts
- Gene Symbol
- AOC4P
- Entrez Gene ID
- 90586
- Gene Name
- amine oxidase, copper containing 4, pseudogene
- Gene Aliases
- AOC4, UPAT
- Chromosome Location
- Chr. 17: 42867145 - 42869217 on Build GRCh38
- UniGene ID
- Hs.367781
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002773.1 |
|
1 |
1536 |
|
GenBank mRNA |
AF047485.1 |
- |
1 |
1536 |
|
AF047486.1 |
- |
2 |
325 |
|
View Full Product Details