Gene Transcripts

Gene Symbol
MIR9-3HG
Entrez Gene ID
254559
Gene Name
MIR9-3 host gene
Gene Aliases
LINC00925
Chromosome Location
Chr. 15: 89361579 - 89398487 on Build GRCh38
UniGene ID
Hs.136313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015411.1 7 3714
NR_133001.1 7 4294
NR_133002.1 6 3595
NR_133003.1 7 3629
GenBank mRNA AK054710.1 - 1 1330
AK096359.1 - 1 1829

View Full Product Details