Gene Transcripts
- Gene Symbol
- CSF2RA
- Entrez Gene ID
- 1438
- Gene Name
- colony stimulating factor 2 receptor alpha subunit
- Gene Aliases
- CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
- Chromosome Location
- Chr. X: 1268800 - 1325097 on Build GRCh38
- UniGene ID
- Hs.520937
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027760.1 |
|
9, 10 |
1001 |
|
GenBank mRNA |
L29348.1 |
- |
9, 10 |
956 |
|
View Full Product Details