Gene Transcripts
- Gene Symbol
- PPBPP2
- Entrez Gene ID
- 10895
- Gene Name
- pro-platelet basic protein pseudogene 2
- Gene Aliases
- PPBPL2, SPBPBP
- Chromosome Location
- Chr. 4: 74054038 - 74055399 on Build GRCh38
- UniGene ID
- Hs.3134
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026769.1 |
|
1 |
1035 |
|
GenBank mRNA |
L10403.1 |
- |
1 |
1035 |
|
View Full Product Details