Gene Transcripts
- Gene Symbol
- LINC00515
- Entrez Gene ID
- 282566
- Gene Name
- long intergenic non-protein coding RNA 515
- Gene Aliases
- C21orf71, PRED21
- Chromosome Location
- Chr. 21: 25582775 - 25583224 on Build GRCh38
- UniGene ID
- Hs.384586
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024092.1 |
|
1 |
422 |
|
GenBank mRNA |
AF086441.1 |
- |
1 |
422 |
|
View Full Product Details