Gene Transcripts
- Gene Symbol
- MRGPRG-AS1
- Entrez Gene ID
- 283303
- Gene Name
- MRGPRG antisense RNA 1
- Gene Aliases
- C11orf36, HSD-40
- Chromosome Location
- Chr. 11: 3218332 - 3223131 on Build GRCh38
- UniGene ID
- Hs.151154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027138.1 |
|
2 |
1530 |
|
GenBank mRNA |
AK093421.1 |
- |
2 |
1529 |
|
AK097749.1 |
- |
3 |
1562 |
|
View Full Product Details