Gene Transcripts
- Gene Symbol
- AKR1C8P
- Entrez Gene ID
- 340811
- Gene Name
- aldo-keto reductase family 1 member C8, pseudogene
- Gene Aliases
- AKR1CL1
- Chromosome Location
- Chr. 10: 5154460 - 5185187 on Build GRCh38
- UniGene ID
- Hs.282513
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027916.2 |
|
8 |
1957 |
|
GenBank mRNA |
AK131335.1 |
- |
8 |
1959 |
|
View Full Product Details