Gene Transcripts
- Gene Symbol
- MEG8
- Entrez Gene ID
- 79104
- Gene Name
- maternally expressed 8 (non-protein coding)
- Gene Aliases
- Bsr, Irm, LINC00024, NCRNA00024, Rian
- Chromosome Location
- Chr. 14: 100894770 - 100906968 on Build GRCh38
- UniGene ID
- Hs.655789
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024149.2 |
|
1 |
222 |
|
GenBank mRNA |
CA396130.1 |
- |
1 |
225 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
T85042.1 |
- |
|
133 |
|
Chr. - - [-] |
View Full Product Details