Gene Transcripts
- Gene Symbol
- DLEU2L
- Entrez Gene ID
- 79469
- Gene Name
- deleted in lymphocytic leukemia 2-like
- Gene Aliases
- BCMSUNL
- Chromosome Location
- Chr. 1: 63548980 - 63550636 on Build GRCh38
- UniGene ID
- Hs.659291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002771.1 |
|
1 |
1460 |
|
GenBank mRNA |
AF254117.1 |
- |
1 |
1460 |
|
- Gene Symbol
- DLEU2
- Entrez Gene ID
- 8847
- Gene Name
- deleted in lymphocytic leukemia 2 (non-protein coding)
- Gene Aliases
- 1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
- Chromosome Location
- Chr. 13: 49982552 - 50125541 on Build GRCh38
- UniGene ID
- Hs.547964
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF264787.1 |
- |
7 |
1821 |
|
View Full Product Details