Gene Transcripts

Gene Symbol
LINC00696
Entrez Gene ID
100128378
Gene Name
long intergenic non-protein coding RNA 696
Gene Aliases
C3orf74
Chromosome Location
Chr. 3: 52062094 - 52065112 on Build GRCh38
UniGene ID
Hs.640106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027331.1 1 1823


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK127958.1 - 1 1823 Chr. 3 - 52062094 - 52065112 [-]

View Full Product Details