Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024430.2 | 4 | 1496 | ||
NR_137175.1 | 2 | 1767 | |||
NR_137176.1 | 2 | 1478 | |||
NR_137177.1 | 2 | 1457 | |||
NR_137179.1 | 5 | 1450 | |||
NR_137180.1 | 5 | 1508 | |||
NR_137181.1 | 3 | 1393 | |||
NR_137182.1 | 4 | 1475 | |||
NR_137183.1 | 3 | 1384 | |||
NR_137184.1 | 2 | 1302 | |||
NR_137185.1 | 3 | 1363 | |||
NR_137186.1 | 3 | 1272 | |||
NR_137187.1 | 3 | 1251 | |||
NR_137188.1 | 3 | 1341 | |||
NR_137189.1 | 2 | 1788 | |||
NR_137190.1 | 3 | 1821 | |||
NR_137191.1 | 3 | 1800 | |||
NR_137192.1 | 4 | 1660 | |||
NR_137193.1 | 5 | 1487 | |||
NR_137194.1 | 3 | 1362 | |||
NR_137195.1 | 2 | 1530 | |||
NR_137196.1 | 2 | 1551 | |||
GenBank mRNA | AB075498.1 | - | 2 | 2020 | |
AK021552.1 | - | 1 | 662 | ||
AK123947.1 | - | 2 | 1704 | ||
AL832469.1 | - | 3 | 1056 | ||
BC040599.1 | - | 2 | 1080 | ||
BX538058.1 | - | 2 | 1058 | ||
BX647608.1 | - | 2 | 1058 |