Gene Transcripts

Gene Symbol
GSTM2P1
Entrez Gene ID
442245
Gene Name
glutathione S-transferase mu 2 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 6: 111046419 - 111047554 on Build GRCh38
UniGene ID
Hs.551627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002932.2 1 497
GenBank mRNA AY532924.1 - 1 464
AY532925.1 - 1 464

View Full Product Details